Naftifine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Naftifine HCl, (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine hydrochloride, AW 105843, SN 105843, N-trans-Cinnamyl-N-methyl-(1-naphthylmethyl)amine hydrochloride

C1162-5G C1162-1G
TCC1162-5GEA 1069.5 CAD
TCC1162-5G TCC1162-001G
Naftifine hydrochloride ≥98.0% (by titrimetric analysis)
Naftifine hydrochloride

Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 177


Other Product Names / Synonyms: Naftifine Hydrochloride

Formula: C₂₁H₂₁N
MW: 323.86 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00059047
CAS Number: 65473-14-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c64bff9566f67bf68d7132a3250fe5f470d4e2bb&vid=c64bff9566f67bf68d7132a3250fe5f470d4e2bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR