Copper(I) trifluoromethanethiolate ≥90.0%

Supplier: TCI America

Synonyms: (Trifluoromethylthio)copper(I), Trifluoromethanethiol Copper(I) Salt

C1159-1G C1159-5G
TCC1159-001GEA 276.5 CAD
TCC1159-001G TCC1159-5G
Copper(I) trifluoromethanethiolate ≥90.0%
Copper(I) trifluoromethanethiolate

CAS Number: 3872-23-9
MDL Number: MFCD00059046
Molecular Formula: CCuF3S
Molecular Weight: 164.61
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis), Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds
Other Product Names / Synonyms: (Trifluoromethylthio)copper(I), Trifluoromethanethiol Copper(I) Salt

Formula: CCuF₃S
MW: 164.61 g/mol
MDL Number: MFCD00059046
CAS Number: 3872-23-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33fe541c01d822e8778048ee456509e0269e8acb&vid=33fe541c01d822e8778048ee456509e0269e8acb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR