Sodium-3-chloro-2-hydroxypropanesulfonate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
C1144-25G
C1144-500G
TCC1144-025GEA
36.34
CAD
TCC1144-025G
TCC1144-500G
Sodium-3-chloro-2-hydroxypropanesulfonate ≥95.0% (by titrimetric analysis)
Sodium-3-chloro-2-hydroxypropanesulfonate
CAS Number: 126-83-0
MDL Number: MFCD00013378
Molecular Formula: C3H7ClO4S
Molecular Weight: 196.58
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Other Product Names / Synonyms: 3-Chloro-2-hydroxypropanesulfonic Acid Sodium Salt
Formula:
C₃H₇ClO₄S |
MDL Number:
MFCD00013378 CAS Number: 126-83-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...