1,2,3-Cyclohexanetriol (cis and trans mixture) ≥97.0%
Supplier: TCI America
Synonyms:
1,2,3-Cyclohexanetriol, Hexahydropyrogallol, Pyrogallitol
C1132-10G
C1132-25G
TCC1132-010GEA
282.82
CAD
TCC1132-010G
TCC1132-025G
1,2,3-Cyclohexanetriol (cis and trans mixture) ≥97.0%
1,2,3-Cyclohexanetriol (cis and trans mixture)
CAS Number: 6286-43-7
MDL Number: MFCD00060111
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Other Product Names / Synonyms: Hexahydropyrogallol, Pyrogallitol
Formula:
C₆H₁₂O₃ |
MDL Number:
MFCD00060111 CAS Number: 6286-43-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...