4-Chlorophenethylamine ≥98.0%
Supplier: TCI America
Synonyms:
2-(4-Chlorophenyl)ethylamine
C1129-25G
C1129-5G
TCC1129-25GEA
123.13
CAD
TCC1129-25G
76219-668
4-Chlorophenethylamine ≥98.0%
4-Chlorophenethylamine
CAS Number: 156-41-2
MDL Number: MFCD00008191
Molecular Formula: C8H10ClN
Molecular Weight: 155.63
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 106
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: 1-Amino-2-(4-chlorophenyl)ethane, 4-Chlorophenethylamine
Formula:
C₈H₁₀ClN Boiling Pt: 140 °C Density: 1.12 (20 °C) Flash Pt: 106 °C |
MDL Number:
MFCD00008191 CAS Number: 156-41-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...