Comanic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1104-5G C1104-25G C1104-1G
TCC1104-5GEA 205.72 CAD
TCC1104-5G TCC1104-25G TCC1104-001G
Comanic acid ≥98.0% (by GC, titration analysis)
Comanic acid

CAS Number: 499-05-8
MDL Number: MFCD00059958
Molecular Formula: C6H4O4
Molecular Weight: 140.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Oxo-4H-pyran-2-carboxylic Acid, gamma-Pyrone-2-carboxylic Acid

Formula: C₆H₄O₄
MDL Number: MFCD00059958
CAS Number: 499-05-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f67fbae55b6d83955644dcca50b3453aa00b62b9&vid=f67fbae55b6d83955644dcca50b3453aa00b62b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR