Ethyl cyclohexylacetate ≥98.0%
Supplier: TCI America
C1076-25ML
C1076-500ML
TCC1076-025MLEA
64.27
CAD
TCC1076-025ML
TCC1076-500ML
Ethyl cyclohexylacetate ≥98.0%
Ethyl cyclohexylacetate
CAS Number: 5452-75-5
MDL Number: MFCD00001517
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 212
Flash Point (°C): 80
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: Cyclohexaneacetic Acid Ethyl Ester, Ethyl Cyclohexaneacetate, Cyclohexylacetic Acid Ethyl Ester
Formula:
C₁₀H₁₈O₂ Boiling Pt: 212 °C Density: 0.95 (20 °C) Flash Pt: 80 °C |
MDL Number:
MFCD00001517 CAS Number: 5452-75-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...