9-Phenanthrenecarbonitrile ≥98.0%

Supplier: TCI America

Synonyms: Phenanthrene-9-carbonitrile, 9-Phenanthrenecarbonitrile, 9-Cyanophenanthrene

C1071-1G C1071-5G
TCC1071-001GEA 151.68 CAD
TCC1071-001G TCC1071-005G
9-Phenanthrenecarbonitrile ≥98.0%
9-Phenanthrenecarbonitrile

CAS Number: 2510-55-6
MDL Number: MFCD00001173
Molecular Formula: C15H9N
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 175
Melting point (°C): 110


Other Product Names / Synonyms: 9-Phenanthrenecarbonitrile

Formula: C₁₅H₉N
MW: 203.24 g/mol
Boiling Pt: 175 °C
Melting Pt: 110 °C
MDL Number: MFCD00001173
CAS Number: 2510-55-6
EINECS: 219-725-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8f218b29e387c1b443ad922f83fda9eb30ba16e&vid=d8f218b29e387c1b443ad922f83fda9eb30ba16e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR