(2-Chloro-1,1-dimethylethyl)benzene ≥98.0%
Supplier: TCI America
Synonyms:
1-Chloro-2-methyl-2-phenylpropane, 2-Chloromethyl-2-phenylpropane
C1068-500ML
C1068-25ML
TCC1068-500MLEA
192.76
CAD
TCC1068-500ML
TCC1068-025ML
(2-Chloro-1,1-dimethylethyl)benzene ≥98.0%
(2-Chloro-1,1-dimethylethyl)benzene
CAS Number: 515-40-2
MDL Number: MFCD00000940
Molecular Formula: C10H13Cl
Molecular Weight: 168.66
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 96
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: 2-Chloromethyl-2-phenylpropane, Neophyl Chloride
Formula:
C₆H₅C(CH₃)₂CH₂Cl MW: 168.67 g/mol Boiling Pt: 222 °C Density: 1.04 (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00000940 CAS Number: 515-40-2 EINECS: 208-197-7 Merck Index: 12,06548 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...