Cystamine sulfate hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C1063-500G
C1063-25G
TCC1063-500GEA
489.8
CAD
TCC1063-500G
TCC1063-025G
Cystamine sulfate hydrate ≥98.0% (by titrimetric analysis)
Cystamine sulfate hydrate
CAS Number: 16214-16-7
MDL Number: MFCD00060254
Molecular Formula: C4H12N2S2
Molecular Weight: 250.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2,2'-Diaminodiethyl Disulfide Sulfate, 2,2'-Dithiobis(ethylamine) Sulfate
Formula:
C₄H₁₂N₂S₂ |
MDL Number:
MFCD00060254 CAS Number: 16214-16-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...