N-Cinnamoyl-N-(2,3-xylyl)hydroxylamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C1058-1G C1058-5G
TCC1058-001GEA 94.8 CAD
TCC1058-001G TCC1058-5G
N-Cinnamoyl-N-(2,3-xylyl)hydroxylamine ≥98.0% (by HPLC, total nitrogen)
N-Cinnamoyl-N-(2,3-xylyl)hydroxylamine

CAS Number: 69891-38-9
MDL Number: MFCD00059308
Molecular Formula: C17H17NO2
Molecular Weight: 267.33
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 152


Product Categories: Analytical Science, Bipyridyls, etc. (Chelating Agents)
Other Product Names / Synonyms: N-(2,3-Xylyl)-N-cinnamoylhydroxylamine

Formula: C₁₇H₁₇NO₂
Melting Pt: 152 °C
MDL Number: MFCD00059308
CAS Number: 69891-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c820c1560774c3739878ffcbabf6a329a3a083a&vid=4c820c1560774c3739878ffcbabf6a329a3a083a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR