Carbazole-9-carbonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 9-(Chlorocarbonyl)carbazole

C1031-1G C1031-5G C1031-25G
TCC1031-001GEA 61.78 CAD
TCC1031-001G TCC1031-5G TCC1031-25G
Carbazole-9-carbonyl chloride ≥98.0% (by GC, titration analysis)
Carbazole-9-carbonyl chloride

CAS Number: 73500-82-0
MDL Number: MFCD00059689
Molecular Formula: C13H8ClNO
Molecular Weight: 229.66
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 105


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 9-(Chlorocarbonyl)carbazole

Formula: C₁₃H₈ClNO
MW: 229.66 g/mol
Melting Pt: 105 °C
MDL Number: MFCD00059689
CAS Number: 73500-82-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b997d5ea1434d5a580ff8994b21bca2ab202653b&vid=b997d5ea1434d5a580ff8994b21bca2ab202653b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR