Propyl cyanoacetate ≥99.0%

Supplier: TCI America

C1016-500ML C1016-25ML
TCC1016-500MLEA 456.62 CAD
TCC1016-500ML TCC1016-025ML
Propyl cyanoacetate ≥99.0%
Propyl cyanoacetate

CAS Number: 14447-15-5
MDL Number: MFCD00015784
Molecular Formula: C6H9NO2
Molecular Weight: 127.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 216
Flash Point (°C): 108
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Cyanoacetic Acid Propyl Ester

Formula: C₆H₉NO₂
Boiling Pt: 216 °C
Density: 1.03 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00015784
CAS Number: 14447-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0f32eda81ac4131f5cddca0f96fadf83e24f6a6&vid=f0f32eda81ac4131f5cddca0f96fadf83e24f6a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR