Isobutyl cyanoacetate ≥98.0%
Supplier: TCI America
C1000-500ML
C1000-25ML
TCC1000-500MLEA
194.34
CAD
TCC1000-500ML
TCC1000-025ML
Isobutyl cyanoacetate ≥98.0%
Isobutyl cyanoacetate
CAS Number: 13361-31-4
MDL Number: MFCD00044320
Molecular Formula: C7H11NO2
Molecular Weight: 141.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 223
Flash Point (°C): 107
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: Cyanoacetic Acid Isobutyl Ester
Formula:
C₇H₁₁NO₂ |
MDL Number:
MFCD00044320 CAS Number: 13361-31-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...