α-Cyclopropylbenzyl alcohol ≥96.0%

Supplier: TCI America

C0986-25ML C0986-5ML
TCC0986-025MLEA 568.8 CAD
TCC0986-025ML TCC0986-5ML
α-Cyclopropylbenzyl alcohol ≥96.0%
α-Cyclopropylbenzyl alcohol

CAS Number: 1007-03-0
MDL Number: MFCD00001299
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 112
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Cyclopropylphenylcarbinol

Formula: C₁₀H₁₂O
Boiling Pt: 132 °C
Density: 1.04 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00001299
CAS Number: 1007-03-0
EINECS: 213-749-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=def6bfdb07e0c93c77e17e941d54667ac15d80e3&vid=def6bfdb07e0c93c77e17e941d54667ac15d80e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR