Chromeazurol S (Mordant blue 29)

Supplier: TCI America

C0979-25G C0979-5G
TCC0979-025GEA 231.52 CAD
TCC0979-025G TCC0979-005G
Chromeazurol S (Mordant blue 29)
Chromeazurol S (Mordant blue 29)

CAS Number: 1667-99-8 MDL Number: MFCD00001615 Molecular Formula: C23H16Cl2O9S Molecular Weight: 605.28 Form: Crystal Color: Deep Yellow Red


Form: Crystal
Color: Deep Yellow Red

Formula: C₂₃H₁₆Cl₂O₉S
MDL Number: MFCD00001615
CAS Number: 1667-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a6a6dca0f7fa55982119f2cee6c65febc2e8830&vid=8a6a6dca0f7fa55982119f2cee6c65febc2e8830&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Lambda max. 425.0 to 435.0 nm(pH7.0 phosphate buffer)
Absorbance min. 0.500(0.02g/L,pH7.0 phosphate buffer,425.0 to 435.0 nm)(calcd.on dried substance)
Drying loss max. 10.0 %
Ignition residue(Sulfate) 35.0 to 45.0% (calcd.on dried substance)
Sensitiveness Colorimetric Test : Turn Purple with Aluminum and turn Yellow red by adding EDTA 2Na


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR