2-Ethylhexyl-2-cyano-3,3-diphenylacrylate ≥98.0%

Supplier: TCI America

Synonyms: Octocrylene2-Cyano-3,3-diphenylacrylic Acid 2-Ethylhexyl EsterOctyl 2-Cyano-3,3-diphenylacrylate2-Ethylhexyl 2-cyano-3,3-diphenylacrylate

C0968-25G C0968-500G C0968-100G
TCC0968-100GEA 160.09 CAD
TCC0968-100G TCC0968-025G TCC0968-500G
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate ≥98.0%
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate

CAS Number: 6197-30-4
MDL Number: MFCD00059260
Molecular Formula: C24H27NO2
Molecular Weight: 361.49
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 218
Melting point (°C): -10
Flash Point (°C): 203
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 2-Cyano-3,3-diphenylacrylic Acid 2-Ethylhexyl Ester, Octyl 2-Cyano-3,3-diphenylacrylate

Formula: C₂₄H₂₇NO₂
MW: 361.49 g/mol
Boiling Pt: 218 °C
Melting Pt: –10 °C
Density: 1.05 (20 °C)
Flash Pt: 203 °C
MDL Number: MFCD00059260
CAS Number: 6197-30-4
https://ca.vwr.com/stibo/web/std.lang.all/82/16/28938216.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR