3-Methoxybutyl acetate ≥99.0%

Supplier: TCI America

A0759-25ML A0759-500ML
TCA0759-025MLEA 57.83 CAD
TCA0759-025ML TCA0759-500ML
3-Methoxybutyl acetate ≥99.0%
3-Methoxybutyl acetate

CAS Number: 4435-53-4
MDL Number: MFCD00043928
Molecular Formula: C7H14O3
Molecular Weight: 146.19
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 62
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Acetic Acid 3-Methoxybutyl Ester

Formula: C₇H₁₄O₃
Boiling Pt: 172 °C
Density: 0.96 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00043928
CAS Number: 4435-53-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96ec73b2f2e4447bb560961553d27090092d7e23&vid=96ec73b2f2e4447bb560961553d27090092d7e23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR