Ethyl-3-cyclohexene-1-carboxylate ≥95.0%
Supplier: TCI America
Synonyms:
3-Cyclohexene-1-carboxylic Acid Ethyl Ester
C0966-25ML
C0966-5ML
TCC0966-025MLEA
336.9
CAD
TCC0966-025ML
TCC0966-005ML
Ethyl-3-cyclohexene-1-carboxylate ≥95.0%
Ethyl-3-cyclohexene-1-carboxylate
CAS Number: 15111-56-5
MDL Number: MFCD00037158
Molecular Formula: C9H14O2
Molecular Weight: 154.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: 3-Cyclohexene-1-carboxylic Acid Ethyl Ester
Formula:
C₉H₁₄O₂ MW: 154.21 g/mol Boiling Pt: 195 °C Density: 0.99 (20 °C) |
MDL Number:
MFCD00037158 CAS Number: 15111-56-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...