Ethyl-2-cyano-3,3-diphenylacrylate ≥98.0%

Supplier: TCI America

Synonyms: 2-Cyano-3,3-diphenylacrylic Acid Ethyl Ester, Ethyl 2-cyano-3,3-diphenylacrylate

C0962-500G C0962-25G
TCC0962-500GEA 327.22 CAD
TCC0962-500G TCC0962-025G
Ethyl-2-cyano-3,3-diphenylacrylate ≥98.0%
Ethyl-2-cyano-3,3-diphenylacrylate

CAS Number: 5232-99-5
MDL Number: MFCD00027364
Molecular Formula: C18H15NO2
Molecular Weight: 277.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 98


Other Product Names / Synonyms: 2-Cyano-3,3-diphenylacrylic Acid Ethyl Ester

Formula: C₁₈H₁₅NO₂
MW: 277.32 g/mol
Boiling Pt: 200 °C
Melting Pt: 98 °C
MDL Number: MFCD00027364
CAS Number: 5232-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6d16de0776d7d9a7b8181cf96fc276c78631b4c&vid=f6d16de0776d7d9a7b8181cf96fc276c78631b4c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR