Chelidonic acid monohydrate ≥95.0%
Supplier: TCI America
Synonyms:
4-Oxo-4H-pyran-2,6-dicarboxylic acid monohydrate, 2,6-Dicarboxy-4-oxo-4H-pyran monohydrate
C0955-25G
C0955-5G
TCC0955-25GEA
627.26
CAD
TCC0955-25G
TCC0955-005G
Chelidonic acid monohydrate ≥95.0%
Chelidonic acid monohydrate
CAS Number: 6003-94-7
MDL Number: MFCD00149433
Molecular Formula: C7H4O6
Molecular Weight: 184.10
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Monomers, Bioscience, Others (Phytochemicals)
Other Product Names / Synonyms: 4-Oxo-4H-pyran-2,6-dicarboxylic Acid
Formula:
C₇H₄O₆.H₂O MW: 202.12 g/mol |
MDL Number:
MFCD00149433 CAS Number: 6003-94-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...