1-(4-Chlorophenyl)-3-methyl-5-pyrazolone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0953-25G C0953-500G
TCC0953-025GEA 47.4 CAD
TCC0953-025G TCC0953-500G
1-(4-Chlorophenyl)-3-methyl-5-pyrazolone ≥98.0% (by titrimetric analysis)
1-(4-Chlorophenyl)-3-methyl-5-pyrazolone

CAS Number: 13024-90-3
MDL Number: MFCD00043814
Molecular Formula: C10H9ClN2O
Molecular Weight: 208.65
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 167

Formula: C₁₀H₉ClN₂O
Melting Pt: 167 °C
MDL Number: MFCD00043814
CAS Number: 13024-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42e08381205f0b4115807b4e0666a8d94de94ec4&vid=42e08381205f0b4115807b4e0666a8d94de94ec4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR