4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine ≥99.0%

Supplier: TCI America

Synonyms: 4-((5-Chloropyridin-2-yl)diazenyl)benzene-1,3-diamine

C0932-1G C0932-100MG
TCC0932-001GEA 352.34 CAD
TCC0932-001G TCC0932-100MG
4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine ≥99.0%
4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine

[for Colorimetric Analysis of Co, Cd]
CAS Number: 33006-91-6
MDL Number: MFCD00059781
Molecular Formula: C11H10ClN5
Molecular Weight: 247.69
Purity/Analysis Method: >99.0% (T)
Form: Crystal

Formula: C₁₁H₁₀ClN₅
MW: 247.69 g/mol
MDL Number: MFCD00059781
CAS Number: 33006-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0420b41fd2c4abaad34be3502cca9fc39a6ccc48&vid=0420b41fd2c4abaad34be3502cca9fc39a6ccc48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR