Ethyl-2-oxocyclohexanecarboxylate ≥95.0%
Supplier: TCI America
C0913-25ML
C0913-500ML
C0913-100ML
TCC0913-025MLEA
72.68
CAD
TCC0913-025ML
TCC0913-500ML
TCC0913-100ML
Ethyl-2-oxocyclohexanecarboxylate ≥95.0%
Ethyl 2-oxocyclohexanecarboxylate
CAS Number: 1655-07-8
MDL Number: MFCD00001631
Molecular Formula: C9H14O3
Molecular Weight: 170.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 85
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 2-Oxocyclohexanecarboxylic Acid Ethyl Ester
Formula:
C₉H₁₄O₃ Boiling Pt: 160 °C Density: 1.07 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00001631 CAS Number: 1655-07-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...