β-Cyclodextrine ≥98.0% (by HPLC)

Supplier: TCI America

C0900-25G C0900-500G C0900-100G
TCC0900-025GEA 57.4 CAD
TCC0900-025G TCC0900-500G TCC0900-100G
β-Cyclodextrine ≥98.0% (by HPLC)
β-Cyclodextrine

CAS Number: 7585-39-9
MDL Number: MFCD00078139
Molecular Formula: C42H70O35
Molecular Weight: 1134.99
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 163 deg (C=1, H2O)


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Oligosaccharides
Other Product Names / Synonyms: Schardinger beta-Dextrin

Formula: C₄₂H₇₀O₃₅
MDL Number: MFCD00078139
CAS Number: 7585-39-9
EINECS: 231-493-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f87dbb3e5ab95b46156c93ca5152f29a440336ee&vid=f87dbb3e5ab95b46156c93ca5152f29a440336ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR