Vinyl crotonate ≥99.0% stabilised

Supplier: TCI America

Synonyms: Crotonic Acid Vinyl Ester

C0897-500ML C0897-25ML
TCC0897-500MLEA 571.96 CAD
TCC0897-500ML TCC0897-025ML
Vinyl crotonate ≥99.0% stabilised
Vinyl crotonate

(stabilized with MEHQ)
CAS Number: 14861-06-4
MDL Number: MFCD00009288
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Melting point (°C): -60
Flash Point (°C): 32
Specific Gravity (20/20): 0.94
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Crotonic Acid Vinyl Ester

Formula: C₆H₈O₂
Boiling Pt: 134 °C
Melting Pt: –60 °C
Density: 0.94 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00009288
CAS Number: 14861-06-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c290c7061c6b77fd041fdb7f3efd2f3d5c74c583&vid=c290c7061c6b77fd041fdb7f3efd2f3d5c74c583&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR