Vinyl-n-octanoate ≥99.0% stabilized
Supplier: TCI America
Synonyms:
n-Octanoic Acid Vinyl Ester
C0896-500ML
C0896-25ML
TCC0896-500MLEA
447.14
CAD
TCC0896-500ML
TCC0896-025ML
Vinyl-n-octanoate ≥99.0% stabilized
Vinyl-n-octanoate
(stabilized with MEHQ)
CAS Number: 818-44-0
MDL Number: MFCD00048915
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 75
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: n-Octanoic Acid Vinyl Ester
Formula:
C₁₀H₁₈O₂ MW: 170.25 g/mol Boiling Pt: 135 °C Density: 0.88 (20 °C) Flash Pt: 75 °C |
MDL Number:
MFCD00048915 CAS Number: 818-44-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...