Cystamine dihydrochloride ≥97.0% for synthesis

Supplier: TCI America

Synonyms: Cystaminium dichloride, 2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Diaminodiethyl disulphide dihydrochloride, 2,2'-Disulphanediyldiethanamine dihydrochloride, Bis(2-aminoethyl) disulphide dihydrochloride, {2-[(2-Aminoethyl)disulfanyl]ethyl}amine dihydrochloride, Decarboxycystine dihydrochloride, Cystamine (dihydrochloride), Bis(2-aminoethyl) disulfide dihydrochloride, {2-[(2-Aminoethyl)disulphanyl]ethyl}amine dihydrochloride, 2,2'-Disulfanediyldiethanamine dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride

C0875-25G C0875-500G C0875-100G
TCC0875-025GEA 78.46 CAD
TCC0875-025G TCC0875-500G TCC0875-100G
Cystamine dihydrochloride ≥97.0% for synthesis
Cystamine dihydrochloride

CAS Number: 56-17-7
MDL Number: MFCD00012905
Molecular Formula: C4H12N2S2
Molecular Weight: 225.19
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 218


Product Categories: Bioscience, Functionality Creating Reagents
Other Product Names / Synonyms: Bis(2-aminoethyl) Disulfide Dihydrochloride, 2,2'-Dithiobis(ethylamine) Dihydrochloride

Formula: C₄H₁₄Cl₂N₂S₂
MW: 225.20 g/mol
Melting Pt: 218 °C
MDL Number: MFCD00012905
CAS Number: 56-17-7
EINECS: 200-260-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=952672efb2c9729c5fef1106664ef96115ca4a34&vid=952672efb2c9729c5fef1106664ef96115ca4a34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR