p-Tolyl crotonate 55.0%

Supplier: TCI America

Synonyms: Crotonic Acid p-Cresyl Ester, Crotonic Acid p-Tolyl Ester, p-Cresyl Crotonate

C0866-500ML C0866-25ML
TCC0866-500MLEA 480.95 CAD
TCC0866-500ML TCC0866-025ML
p-Tolyl crotonate 55.0%
p-Tolyl crotonate

CAS Number: 41873-74-9
MDL Number: MFCD00059385
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >55.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: p-Cresyl Crotonate, Crotonic Acid p-Tolyl Ester, Crotonic Acid p-Cresyl Ester

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 158 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00059385
CAS Number: 41873-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec7d5f13fdf95c7ec8efa8882cd610058a9b2f8d&vid=ec7d5f13fdf95c7ec8efa8882cd610058a9b2f8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 55.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR