1,2,3,4-Cyclopentanetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0857-25G C0857-5G
TCC0857-025GEA 851.3 CAD
TCC0857-025G TCC0857-005G
1,2,3,4-Cyclopentanetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis)
1,2,3,4-Cyclopentanetetracarboxylic dianhydride

CAS Number: 6053-68-5
MDL Number: MFCD00046872
Molecular Formula: C9H6O6
Molecular Weight: 210.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 213

Formula: C₉H₆O₆
Melting Pt: 213 °C
MDL Number: MFCD00046872
CAS Number: 6053-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e28f5ee82d3eafb6668bae1b55716a610b7f54da&vid=e28f5ee82d3eafb6668bae1b55716a610b7f54da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR