N-Acetyl-DL-norleucine ≥98.0%

Supplier: TCI America

Synonyms: Ac-DL-Nle-OH

A0750-1G A0750-25G
TCA0750-001GEA 37.92 CAD
TCA0750-001G TCA0750-025G
N-Acetyl-DL-norleucine ≥98.0%
N-Acetyl-DL-norleucine

CAS Number: 7682-16-8
MDL Number: MFCD00037272
Molecular Formula: C8H15NO3
Molecular Weight: 173.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 108


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-DL-Nle-OH

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00037272
CAS Number: 7682-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cfab0218278b56e1daa2a38457f09a5da46fc3a&vid=3cfab0218278b56e1daa2a38457f09a5da46fc3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR