1,2,3,4-Cyclopentanetetracarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C0856-25G C0856-5G
TCC0856-025GEA 274.45 CAD
TCC0856-025G 76219-658
1,2,3,4-Cyclopentanetetracarboxylic acid ≥98.0% (by GC, titration analysis)
1,2,3,4-Cyclopentanetetracarboxylic acid

CAS Number: 3724-52-5
MDL Number: MFCD00001377
Molecular Formula: C9H10O8
Molecular Weight: 246.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₉H₁₀O₈
MDL Number: MFCD00001377
CAS Number: 3724-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a48c4574714d264a291280e867b25fdec3d1a921&vid=a48c4574714d264a291280e867b25fdec3d1a921&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR