3-Carboxy-1-(4-sulfophenyl)-5-pyrazolone sodium salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C0845-500G C0845-25G
TCC0845-500GEA 712.58 CAD
TCC0845-500G TCC0845-025G
3-Carboxy-1-(4-sulfophenyl)-5-pyrazolone sodium salt ≥98.0% (by HPLC, titration analysis)
3-Carboxy-1-(4-sulfophenyl)-5-pyrazolone sodium salt

CAS Number: 52126-51-9
MDL Number: MFCD00059720
Molecular Formula: C10H8N2O6S
Molecular Weight: 306.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 5-Oxo-1-(4-sulfophenyl)-2-pyrazoline-3-carboxylic Acid Sodium Salt

Formula: C₁₀H₈N₂O₆S
MDL Number: MFCD00059720
CAS Number: 52126-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76fe4f5c53215655162d2873c210468b27886c17&vid=96903bf3ea5006b3fa0dadac6b17f9b4eed06fee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR