3-Aminocyclohexanecarboxylic acid (cis and trans mixture) ≥95.0% (by titrimetric analysis)
Supplier: TCI America
A0749-5G
A0749-25G
TCA0749-005GEA
128.93
CAD
TCA0749-005G
TCA0749-025G
3-Aminocyclohexanecarboxylic acid (cis and trans mixture) ≥95.0% (by titrimetric analysis)
3-Aminocyclohexanecarboxylic acid (cis and trans mixture)
CAS Number: 25912-50-9
MDL Number: MFCD00059562
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Formula:
C₇H₁₃NO₂ |
MDL Number:
MFCD00059562 CAS Number: 25912-50-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...