Chitosan, 200-600 mPa.s (20 °C) 0.5% in acetic acid (0,5%)
Supplier: TCI America
Synonyms:
Deacetylated chitin, Poly(D-glucosamine)
C0831-500G
C0831-25G
TCC0831-500GEA
316.55
CAD
TCC0831-500G
TCC0831-025G
Chitosan, 200-600 mPa.s (20 °C) 0.5% in acetic acid (0,5%)
Chitosan
CAS Number: 9012-76-4
MDL Number: MFCD00161512
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Natural Polymers (for DDS)
MDL Number:
MFCD00161512 CAS Number: 9012-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...