β-Caryophyllene ≥90.0%

Supplier: TCI America

C0796-25ML C0796-500ML C0796-100ML
TCC0796-025MLEA 85.16 CAD
TCC0796-025ML TCC0796-500ML TCC0796-100ML
β-Caryophyllene ≥90.0%
β-Caryophyllene

CAS Number: 87-44-5
MDL Number: MFCD00075925
Molecular Formula: C15H24
Molecular Weight: 204.36
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 97
Specific Gravity (20/20): 0.90
Specific rotation [a]20/D: -9.2 deg (neat)

Formula: C₁₅H₂₄
Boiling Pt: 116 °C
Density: 0.90 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00075925
CAS Number: 87-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a5fc40d1744723216758b01d45a1112c00ca128&vid=9a5fc40d1744723216758b01d45a1112c00ca128&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR