1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulfonate ≥96.0%
Supplier: TCI America
Synonyms:
CMC
C0793-25G
C0793-5G
TCC0793-25GEA
2109.3
CAD
TCC0793-25G
TCC0793-005G
1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulfonate ≥96.0%
1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulfonate
[for Peptide Synthesis]
CAS Number: 2491-17-0
MDL Number: MFCD00011979
Molecular Formula: C21H33N3O4S
Molecular Weight: 423.57
Purity/Analysis Method: >96.0% (N,T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Bioscience, Crosslinkers
Other Product Names / Synonyms: CMC
Formula:
C₂₁H₃₃N₃O₄S MW: 423.57 g/mol |
MDL Number:
MFCD00011979 CAS Number: 2491-17-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...