cis-1,4-Cyclohexanedicarboxylic acid ≥90.0%
Supplier: TCI America
Synonyms:
cis-Cyclohexane-1,4-dicarboxylic acid
C0789-25G
C0789-100G
TCC0789-025GEA
151.02
CAD
TCC0789-025G
TCC0789-100G
cis-1,4-Cyclohexanedicarboxylic acid ≥90.0%
cis-1,4-Cyclohexanedicarboxylic acid
CAS Number: 619-81-8
MDL Number: MFCD00064950
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Melting point (°C): 168
Formula:
C₈H₁₂O₄ Melting Pt: 168 °C |
MDL Number:
MFCD00064950 CAS Number: 619-81-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...