Carminic acid (natural)

Supplier: TCI America

C0782-25G C0782-5G
TCC0782-025GEA 674.66 CAD
TCC0782-025G TCC0782-005G
Carminic acid (natural)
Carminic acid

CAS Number: 1260-17-9
MDL Number: MFCD00167028
Molecular Formula: C22H20O13
Molecular Weight: 492.39
Form: Crystal
Color: Yellow Red

Formula: C₂₂H₂₀O₁₃
MDL Number: MFCD00167028
CAS Number: 1260-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6860bdaa7574842f21fa1c07578b1de5f2ed412a&vid=af3b4a6fd6327c796cdb91cbaaf952fce34c4305&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR