β-Cyclodextrine ≥99.0% (by HPLC)

Supplier: TCI America

C0777-25G C0777-100G
TCC0777-025GEA 157.53 CAD
TCC0777-025G TCC0777-100G
β-Cyclodextrine ≥99.0% (by HPLC)
β-Cyclodextrine

CAS Number: 7585-39-9
MDL Number: MFCD00078139
Molecular Formula: C42H70O35
Molecular Weight: 1134.99
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 163 deg (C=1, H2O)


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Oligosaccharides, Cyclodextrins
Other Product Names / Synonyms: Cycloheptaamylose, Schardinger beta-Dextrin

Formula: C₄₂H₇₀O₃₅
MDL Number: MFCD00078139
CAS Number: 7585-39-9
EINECS: 231-493-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=540ac9b8f0bfb0fbe7b89cb6e9c00987c1a1e39e&vid=540ac9b8f0bfb0fbe7b89cb6e9c00987c1a1e39e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR