Butyl-4-Carboxyphenyl carbonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0773-1G C0773-5G
TCC0773-001GEA 175.38 CAD
TCC0773-001G TCC0773-5G
Butyl-4-Carboxyphenyl carbonate ≥98.0% (by titrimetric analysis)
Butyl-4-Carboxyphenyl carbonate

CAS Number: 14180-12-2
MDL Number: MFCD00060074
Molecular Formula: C12H14O5
Molecular Weight: 238.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 146


Other Product Names / Synonyms: 4-Carboxyphenyl Butyl Carbonate

Formula: C₁₂H₁₄O₅
Melting Pt: 146 °C
MDL Number: MFCD00060074
CAS Number: 14180-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b09d6b5f52763b52c892ba851b4745e7c266a61&vid=5b09d6b5f52763b52c892ba851b4745e7c266a61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR