N-Acetyl-D-phenylalanine ≥98.0%
Supplier: TCI America
Synonyms:
N-Ac-D-Phe-OH, Ac-D-Phe-OH, N-Acetyl-D-Phenylalanine
A0105-1G
A0105-5G
TCA0105-001GEA
42.66
CAD
TCA0105-001G
TCA0105-5G
N-Acetyl-D-phenylalanine ≥98.0%
N-Acetyl-D-phenylalanine
CAS Number: 10172-89-1
MDL Number: MFCD00002664
Molecular Formula: C11H13NO3
Molecular Weight: 207.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 167
Specific rotation [a]20/D: -41 deg (C=5, MeOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-D-Phe-OH
Formula:
C₁₁H₁₃NO₃ Melting Pt: 167 °C |
MDL Number:
MFCD00002664 CAS Number: 10172-89-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...