2-Cinnamoylthiophene 98,0

Supplier: TCI America

Synonyms: Styryl 2-Thienyl Ketone, 3-Phenyl-1-(thiophen-2-yl)prop-2-en-1-one, 3-Phenyl-1-(2-thienyl)-2-propen-1-one

C0755-5G C0755-1G
TCC0755-5GEA 289.3 CAD
TCC0755-5G TCC0755-001G
2-Cinnamoylthiophene 98,0
2-Cinnamoylthiophene

CAS Number: 3988-77-0
MDL Number: MFCD00059742
Molecular Formula: C13H10OS
Molecular Weight: 214.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82


Other Product Names / Synonyms: 3-Phenyl-1-(2-thienyl)-2-propen-1-one, Styryl 2-Thienyl Ketone

Formula: C₁₃H₁₀OS
MW: 214.28 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00059742
CAS Number: 3988-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94fc6d88d02c4e00b93bd4a62ea496264c008ec8&vid=94fc6d88d02c4e00b93bd4a62ea496264c008ec8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR