N-Carbobenzoxy-4-aminobutyric acid ≥98.0%
Supplier: TCI America
Synonyms:
N-Carbobenzoxy-4-aminobutyric acid, N-Cbz-4-aminobutanoic acid, 4-(Benzyloxycarbonylamino)butyric acid, Z-GABA-OH, N-Cbz-4-aminobutyric acid, Z-γ-Abu-OH
CAS Number: 5105-78-2
MDL Number: MFCD00055835
Molecular Formula: C12H15NO4
Molecular Weight: 237.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 64
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-4-aminobutyric Acid, Z-GABA-OH
Formula:
C₁₂H₁₅NO₄ Melting Pt: 64 °C |
MDL Number:
MFCD00055835 CAS Number: 5105-78-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...