N-Carbobenzoxy-DL-norvaline 98,0

Supplier: TCI America

Synonyms: Z-DL-Nva-OH, N-Benzyloxycarbonyl-DL-norvaline, N-Cbz-DL-norvaline

C0752-5G C0752-25G
TCC0752-005GEA 48.98 CAD
TCC0752-005G TCC0752-025G
N-Carbobenzoxy-DL-norvaline 98,0
N-Carbobenzoxy-DL-norvaline

CAS Number: 21691-43-0
MDL Number: MFCD00020434
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 88


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-DL-norvaline, Z-DL-Nva-OH

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Melting Pt: 88 °C
MDL Number: MFCD00020434
CAS Number: 21691-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba9f1aa5580ab3186bb5a59798d9df46c2cfc96b&vid=ba9f1aa5580ab3186bb5a59798d9df46c2cfc96b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR