(+)-Catechin hydrate ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
(+)-Catechin, (+)-3,3',4',5,7-Flavanpentol hydrate, (2R,3S)-2-(3,4-Dihydroxyphenyl)chromane-3,5,7-triol
CAS Number: 154-23-4
MDL Number: MFCD00075649
Molecular Formula: C15H14O6
Molecular Weight: 290.27
Purity/Analysis Method:
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Specific rotation [a]20/D: 16 deg (C=5, Acetone(50%))
Formula:
C₁₅H₁₄O₆·xH₂O Melting Pt: 96 °C |
MDL Number:
MFCD00075649 CAS Number: 154-23-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...