Coomassie® brilliant blue R-250
Supplier: TCI America
Synonyms:
Brillant blue R
, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-benzenemethanaminium inner salt, sodium salt (1:1)
, Brilliant Blue R250
, Brilliant indocyanine 6B
, Brilliant Blue R
, Serva Blue R
, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulphophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulpho-benzenemethanaminium inner salt, sodium salt (1:1)
, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium salt
, Coomassie Blue R 250
, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt
, Acid Blue 83
, Brilliant Cyanine 6B
CAS Number: 6104-59-2
MDL Number: MFCD00041762
Molecular Formula: C45H45N3O7S2
Molecular Weight: 825.97
Form: Crystal
Color: Purple
Other Product Names / Synonyms: Acid Cyanine 6B, Alizarin Rubinol 5G, Solar Cyanine 6B, Acid Blue 83
Formula:
C₄₅H₄₄N₃NaO₇S₂ MW: 825.98 g/mol Melting Pt: 174…180 °C |
MDL Number:
MFCD00041762 CAS Number: 6104-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...