(R)-2-(((Benzyloxy)carbonyl)amino)succinic acid ≥98.0%

Supplier: TCI America

Synonyms: Cbz-D-Asp-OH, Z-D-Asp-OH, N-Cbz-D-aspartic Acid, N-Benzyloxycarbonyl-D-aspartic acid

C0689-1G C0689-5G
TCC0689-001GEA 26.86 CAD
TCC0689-001G TCC0689-5G
(R)-2-(((Benzyloxy)carbonyl)amino)succinic acid ≥98.0%
(R)-2-(((Benzyloxy)carbonyl)amino)succinic acid

CAS Number: 78663-07-7
MDL Number: MFCD00063182
Molecular Formula: C12H13NO6
Molecular Weight: 267.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 117
Specific rotation [a]20/D: -10 deg (C=7, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-aspartic Acid, Z-D-Asp-OH

Formula: C₁₂H₁₃NO₆
MW: 267.2 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00063182
CAS Number: 78663-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad171527d242f2ccd5c34b0175155726379efe3f&vid=ad171527d242f2ccd5c34b0175155726379efe3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR