Calcium alginate

Supplier: TCI America

A0738-500G A0738-25G
TCA0738-500GEA 252.29 CAD
TCA0738-500G TCA0738-025G
Calcium alginate
Calcium alginate

Form: Crystal
Color: White


Other Product Names / Synonyms: Alginic Acid Calcium Salt

Storage Temperature: Ambient
MDL Number: MFCD00143567
CAS Number: 9005-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR