Nɑ-Carbobenzoxy-D-asparagine 99,0

Supplier: TCI America

Synonyms: Nα-Benzyloxycarbonyl-D-asparagine, Nɑ-Cbz-D-asparagine, Z-D-Asn-OH, Cbz-D-Asn-OH, N-Cbz-D-Asparagine

C0666-100MG C0666-1G
TCC0666-100MGEA 47.4 CAD
TCC0666-100MG TCC0666-001G
Nɑ-Carbobenzoxy-D-asparagine 99,0
Nalpha-Carbobenzoxy-D-asparagine

CAS Number: 4474-86-6
MDL Number: MFCD00065696
Molecular Formula: C12H14N2O5
Molecular Weight: 266.25
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 165
Specific rotation [a]20/D: -6.5 deg (C=1.6, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Cbz-D-asparagine, Z-D-Asn-OH

Formula: C₁₂H₁₄N₂O₅
MW: 266.25 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00065696
CAS Number: 4474-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99e189c8fdb8aec90482ab471fe0a0745c2fb798&vid=99e189c8fdb8aec90482ab471fe0a0745c2fb798&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR